Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024979.1 | NP_001020150.1 | 6 - 7 | 549 | 69 | |
XM_008761129.1 | XP_008759351.1 | 7 - 8 | 1080 | 69 | ||
XM_008761130.1 | XP_008759352.1 | 7 - 8 | 1130 | 69 | ||
XM_008761131.1 | XP_008759353.1 | 6 - 7 | 1066 | 69 | ||
XM_008761132.1 | XP_008759354.1 | 7 - 8 | 1244 | 69 | ||
GenBank mRNA | BC093381.1 | - | 6 - 7 | 549 | 7388663 | 69 |
FQ233538.1 | - | 3 - 4 | 271 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |