Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761239.1 | XP_008759461.1 | 6 - 7 | 1059 | 72 | |
XM_008761240.1 | XP_008759462.1 | 6 - 7 | 1059 | 72 | ||
XM_008761241.1 | XP_008759463.1 | 6 - 7 | 984 | 72 | ||
XM_008761242.1 | XP_008759464.1 | 6 - 7 | 951 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |