Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106439.1 | NP_001099909.1 | 4 - 5 | 1264 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |