Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057140.1 | NP_476481.1 | 9 - 10 | 1128 | 104 | |
GenBank mRNA | BC083859.1 | - | 9 - 10 | 1128 | 7126254 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons |