Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006227982.2 | XP_006228044.1 | 3 - 4 | 405 | 59 | |
XM_006227983.2 | XP_006228045.1 | 3 - 4 | 452 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |