Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181371.2 | NP_852036.1 | 5 - 6 | 455 | 64 | |
XM_006236391.2 | XP_006236453.1 | 4 - 5 | 397 | 64 | ||
GenBank mRNA | FJ179400.1 | - | 5 - 6 | 421 | 64 | |
FQ211030.1 | - | 5 - 6 | 473 | 64 | ||
FQ214179.1 | - | 5 - 6 | 538 | 64 | ||
FQ220456.1 | - | 5 - 6 | 511 | 64 | ||
S83436.1 | - | 5 - 6 | 455 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |