Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181371.2 | NP_852036.1 | 7 - 8 | 668 | 86 | |
XM_006236391.2 | XP_006236453.1 | 6 - 7 | 610 | 86 | ||
GenBank mRNA | FQ211030.1 | - | 7 - 8 | 686 | 86 | |
FQ214179.1 | - | 7 - 8 | 751 | 86 | ||
FQ220456.1 | - | 7 - 8 | 724 | 86 | ||
S83436.1 | - | 7 - 8 | 668 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |