Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004233.1 | NP_001004233.1 | 3 - 4 | 550 | 112 | |
XM_006236301.2 | XP_006236363.1 | 3 - 4 | 750 | 112 | ||
XM_006236302.2 | XP_006236364.1 | 2 - 3 | 622 | 112 | ||
XM_006236303.2 | XP_006236365.1 | 3 - 4 | 706 | 112 | ||
XM_006236304.2 | XP_006236366.1 | 3 - 4 | 566 | 112 | ||
GenBank mRNA | BC079407.1 | - | 3 - 4 | 550 | 7112867 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |