Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cftr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031506.1 | NP_113694.1 | 16 - 17 | 2782 | 105 | |
XM_006236097.2 | XP_006236159.1 | 17 - 18 | 2901 | 105 | ||
XM_008762680.1 | XP_008760902.1 | 17 - 18 | 2901 | 105 | ||
GenBank mRNA | FJ959076.1 | - | 16 - 17 | 2540 | 105 | |
M89906.1 | - | 6 - 7 | 1202 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Inventoried Probe spans exons |