Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cftr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031506.1 | NP_113694.1 | 22 - 23 | 3848 | 133 | |
XM_006236097.2 | XP_006236159.1 | 23 - 24 | 3967 | 133 | ||
GenBank mRNA | FJ959076.1 | - | 22 - 23 | 3606 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |