Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cftr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031506.1 | NP_113694.1 | 25 - 26 | 4255 | 132 | |
XM_006236097.2 | XP_006236159.1 | 26 - 27 | 4374 | 132 | ||
GenBank mRNA | FJ959076.1 | - | 25 - 26 | 4013 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |