Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047863.1 | NP_001041328.1 | 1 - 2 | 224 | 77 | |
XM_006236305.2 | XP_006236367.1 | 2 - 3 | 337 | 77 | ||
GenBank mRNA | DQ480753.1 | - | 1 - 2 | 224 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |