Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100964.1 | NP_001094434.1 | 1 - 2 | 419 | 101 | |
GenBank mRNA | AB331240.1 | - | 1 - 2 | 295 | 101 | |
BC128755.1 | - | 1 - 2 | 419 | 7104774 | 101 |
Gene Symbol: LOC100909784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749707.3 | XP_003749755.1 | 1 - 2 | 491 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |