Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024326.1 | NP_001019497.1 | 1 - 2 | 90 | 114 | |
GenBank mRNA | BC088443.1 | - | 1 - 2 | 90 | 7126386 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |