Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247201.2 | XP_006247263.1 | 1 - 2 | 1119 | 77 | |
XM_006247203.2 | XP_006247265.1 | 1 - 2 | 1119 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |