Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247201.2 | XP_006247263.1 | 3 - 4 | 1921 | 65 | |
XM_006247202.2 | XP_006247264.1 | 3 - 4 | 1886 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |