Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012747.2 | NP_036879.1 | 20 - 21 | 2095 | 77 | |
XM_006247257.2 | XP_006247319.1 | 20 - 21 | 2119 | 77 | ||
XM_006247258.2 | XP_006247320.1 | 20 - 21 | 2119 | 77 | ||
XM_006247259.2 | XP_006247321.1 | 20 - 21 | 2119 | 77 | ||
GenBank mRNA | BC087025.1 | - | 20 - 21 | 2095 | 7110655 | 77 |
FQ223504.1 | - | 1 - 2 | 58 | 77 | ||
X91810.1 | - | 19 - 20 | 1889 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |