Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012747.2 | NP_036879.1 | 2 - 3 | 335 | 61 | |
XM_006247257.2 | XP_006247319.1 | 2 - 3 | 359 | 61 | ||
XM_006247258.2 | XP_006247320.1 | 2 - 3 | 359 | 61 | ||
XM_006247259.2 | XP_006247321.1 | 2 - 3 | 359 | 61 | ||
GenBank mRNA | BC087025.1 | - | 2 - 3 | 335 | 7110655 | 61 |
X91810.1 | - | 1 - 2 | 129 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |