Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053992.1 | NP_446444.1 | 6 - 7 | 1586 | 58 | |
GenBank mRNA | U41662.1 | - | 6 - 7 | 1586 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |