Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109035.1 | NP_001102505.1 | 5 - 6 | 811 | 136 | |
XM_006246786.2 | XP_006246848.1 | 6 - 7 | 1400 | 136 | ||
XM_008767857.1 | XP_008766079.1 | 6 - 7 | 1401 | 136 | ||
XM_008767858.1 | XP_008766080.1 | 6 - 7 | 1401 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |