Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107002.1 | NP_001100472.1 | 8 - 9 | 1208 | 66 | |
XM_006246265.2 | XP_006246327.1 | 8 - 9 | 1420 | 66 | ||
XM_006246266.2 | XP_006246328.1 | 8 - 9 | 1221 | 66 | ||
XM_006246267.2 | XP_006246329.1 | 8 - 9 | 1215 | 66 | ||
XM_006246268.1 | XP_006246330.1 | 8 - 9 | 1183 | 66 | ||
XM_006246270.2 | XP_006246332.1 | 8 - 9 | 1221 | 66 | ||
XM_006246271.2 | XP_006246333.1 | 8 - 9 | 1221 | 66 | ||
GenBank mRNA | BC158796.1 | - | 8 - 9 | 1184 | 7314654 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |