Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013974.1 | NP_001013996.1 | 8 - 9 | 1132 | 156 | |
GenBank mRNA | BC085824.1 | - | 8 - 9 | 1132 | 7127239 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |