Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011911.1 | NP_001011911.1 | 6 - 7 | 999 | 87 | |
XM_006251087.1 | XP_006251149.1 | 6 - 7 | 1005 | 87 | ||
GenBank mRNA | BC079008.1 | - | 6 - 7 | 999 | 7113133 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |