Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130062.1 | NP_001123534.1 | 3 - 4 | 490 | 81 | |
XM_006251239.2 | XP_006251301.1 | 4 - 5 | 1307 | 81 | ||
XM_008770283.1 | XP_008768505.1 | 4 - 5 | 1307 | 81 | ||
GenBank mRNA | FQ223777.1 | - | 4 - 5 | 609 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |