Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130062.1 | NP_001123534.1 | 7 - 8 | 1497 | 78 | |
XM_006251239.2 | XP_006251301.1 | 8 - 9 | 2314 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |