Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdye4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034153.1 | NP_001029325.1 | 1 - 2 | 240 | 85 | |
GenBank mRNA | BC105855.1 | - | 1 - 2 | 240 | 7453404 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |