Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017068.2 | NP_058764.2 | 4 - 5 | 650 | 77 | |
XM_006257486.2 | XP_006257548.1 | 4 - 5 | 681 | 77 | ||
GenBank mRNA | BC061990.1 | - | 4 - 5 | 650 | 5599402 | 77 |
D90211.1 | - | 4 - 5 | 634 | 77 | ||
FQ211284.1 | - | 4 - 5 | 649 | 77 | ||
M32016.1 | - | 3 - 4 | 488 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |