Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191735.1 | NP_001178664.1 | 6 - 7 | 795 | 115 | |
XM_008773515.1 | XP_008771737.1 | 6 - 7 | 3230 | 115 | ||
XM_008773516.1 | XP_008771738.1 | 7 - 8 | 1063 | 115 | ||
XM_008773517.1 | XP_008771739.1 | 7 - 8 | 1261 | 115 | ||
XM_008773518.1 | XP_008771740.1 | 7 - 8 | 2990 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |