Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773415.1 | XP_008771637.1 | 1 - 2 | 68 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |