Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003752132.3 | XP_003752180.2 | 2 - 3 | 1050 | 145 | |
XM_008773488.1 | XP_008771710.1 | 2 - 3 | 1202 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |