Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs6st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257578.2 | XP_006257640.1 | 2 - 3 | 1042 | 84 | |
GenBank mRNA | FQ226114.1 | - | 2 - 3 | 694 | 84 | |
FQ230487.1 | - | 2 - 3 | 694 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |