Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025643.1 | NP_001020814.1 | 1 - 2 | 106 | 64 | |
GenBank mRNA | BC099198.1 | - | 1 - 2 | 106 | 7383920 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |