Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001053214.4 | XP_001053214.2 | 3 - 4 | 461 | 72 | |
XM_006228482.1 | XP_006228544.1 | 3 - 4 | 633 | 72 | ||
XM_006228483.2 | XP_006228545.1 | 4 - 5 | 1170 | 72 | ||
XM_006228484.2 | XP_006228546.1 | 4 - 5 | 524 | 72 | ||
XM_006228485.2 | XP_006228547.1 | 4 - 5 | 853 | 72 | ||
XM_006228486.2 | XP_006228548.1 | 3 - 4 | 1025 | 72 | ||
XM_006228487.1 | XP_006228549.1 | 3 - 4 | 384 | 72 | ||
XM_006228488.2 | XP_006228550.1 | 4 - 5 | 736 | 72 | ||
XM_006228489.1 | XP_006228551.1 | 4 - 5 | 523 | 72 | ||
XM_006228491.2 | XP_006228553.1 | 4 - 5 | 647 | 72 | ||
XM_006228493.1 | XP_006228555.1 | 3 - 4 | 633 | 72 | ||
XM_008759031.1 | XP_008757253.1 | 3 - 4 | 578 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |