Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100821.1 | NP_001094291.1 | 9 - 10 | 1012 | 74 | |
XM_006228904.2 | XP_006228966.1 | 9 - 10 | 1164 | 74 | ||
GenBank mRNA | BC162008.1 | - | 9 - 10 | 1039 | 9090267 | 74 |
D14015.1 | - | 9 - 10 | 1029 | 74 | ||
DY561075.1 | - | 4 - 5 | 448 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |