Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Relb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228502.2 | XP_006228564.1 | 2 - 3 | 288 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Probe spans exons |