Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ffar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228821.2 | XP_006228883.1 | 1 - 2 | 748 | 79 | |
XM_006228822.2 | XP_006228884.1 | 2 - 3 | 342 | 79 | ||
XM_008759133.1 | XP_008757355.1 | 2 - 3 | 442 | 79 | ||
XM_008759134.1 | XP_008757356.1 | 2 - 3 | 403 | 79 | ||
XM_008759135.1 | XP_008757357.1 | 2 - 3 | 120 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |