Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229101.2 | XP_006229163.1 | 9 - 10 | 885 | 73 | |
XM_006229102.2 | XP_006229164.1 | 9 - 10 | 807 | 73 | ||
GenBank mRNA | BC167051.1 | - | 9 - 10 | 723 | 9089731 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |