Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013064.2 | NP_001013082.2 | 2 | 166 | 65 | |
GenBank mRNA | BC079218.1 | - | 2 | 166 | 7131902 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |