Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175765.2 | NP_786941.1 | 12 - 13 | 1297 | 135 | |
XM_006238353.2 | XP_006238415.1 | 13 - 14 | 1524 | 135 | ||
XM_006238354.2 | XP_006238416.1 | 13 - 14 | 1520 | 135 | ||
GenBank mRNA | AB285525.1 | - | 1200 | 135 | ||
AF339084.1 | - | 12 - 13 | 1297 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |