Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750385.3 | XP_003750433.1 | 9 - 10 | 6898 | 62 | |
XM_006242130.2 | XP_006242192.1 | 10 - 11 | 1955 | 62 | ||
XM_008765806.1 | XP_008764028.1 | 9 - 10 | 2568 | 62 | ||
XM_235497.8 | XP_235497.6 | 10 - 11 | 1629 | 62 | ||
GenBank mRNA | AB588919.1 | - | 10 - 11 | 1516 | 62 | |
AB588920.1 | - | 9 - 10 | 1371 | 62 | ||
AB588921.1 | - | 10 - 11 | 1498 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |