Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164060.1 | NP_001157532.1 | 4 - 5 | 955 | 82 | |
NM_053517.2 | NP_445969.2 | 5 - 6 | 522 | 82 | ||
GenBank mRNA | BC088298.1 | - | 5 - 6 | 522 | 7323045 | 82 |
D83015.1 | - | 4 - 5 | 303 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |