Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002016.2 | NP_001002016.2 | 3 - 4 | 843 | 66 | |
GenBank mRNA | BC062018.1 | - | 2 - 3 | 398 | 5621637 | 66 |
DN934590.1 | - | 3 - 4 | 583 | 66 | ||
X66870.1 | - | 3 - 4 | 802 | 66 | ||
X76297.1 | - | 3 - 4 | 565 | 66 | ||
X99257.1 | - | 3 - 4 | 302 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |