Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021589.1 | NP_067600.1 | 7 - 8 | 924 | 63 | |
XM_006232746.2 | XP_006232808.1 | 7 - 8 | 915 | 63 | ||
GenBank mRNA | M85214.1 | - | 7 - 8 | 924 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Rattus norvegicus Probe spans exons |