Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hapln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022285.1 | NP_071621.1 | 3 - 4 | 285 | 93 | |
XM_006232751.2 | XP_006232813.1 | 3 - 4 | 367 | 93 | ||
XM_008761208.1 | XP_008759430.1 | 4 - 5 | 408 | 93 | ||
XM_008761209.1 | XP_008759431.1 | 3 - 4 | 593 | 93 | ||
GenBank mRNA | AB049056.1 | - | 3 - 4 | 285 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |