Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cecr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191977.1 | NP_001178906.1 | 17 - 18 | 4193 | 62 | |
XM_008763251.1 | XP_008761473.1 | 17 - 18 | 4553 | 62 | ||
XM_008763252.1 | XP_008761474.1 | 17 - 18 | 4550 | 62 | ||
XM_008763253.1 | XP_008761475.1 | 16 - 17 | 4469 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |