Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109393.1 | NP_001102863.1 | 1 - 2 | 501 | 133 | |
GenBank mRNA | FQ213131.1 | - | 1 - 2 | 238 | 133 | |
FQ214060.1 | - | 1 - 2 | 209 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |