Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109353.1 | NP_001102823.1 | 2 - 3 | 198 | 111 | |
XM_006237092.2 | XP_006237154.1 | 4 - 5 | 303 | 111 | ||
GenBank mRNA | GU357483.1 | - | 2 - 3 | 185 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |