Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109033.2 | NP_001102503.1 | 1 - 2 | 777 | 158 | |
XM_006246776.2 | XP_006246838.1 | 2 - 3 | 1676 | 158 | ||
XM_008767852.1 | XP_008766074.1 | 1 - 2 | 2099 | 158 | ||
GenBank mRNA | BC158839.1 | - | 1 - 2 | 777 | 7387741 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |