Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100648.1 | NP_001094118.1 | 7 - 8 | 1638 | 67 | |
GenBank mRNA | S80120.1 | - | 7 - 8 | 1638 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |