Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007804.1 | NP_001007805.1 | 4 - 5 | 353 | 89 | |
XM_006247185.1 | XP_006247247.1 | 4 - 5 | 486 | 89 | ||
GenBank mRNA | BC086364.1 | - | 4 - 5 | 396 | 7304427 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |